2-Bromo-4-fluorofenol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
B1555-25G
B1555-5G
TCIAB1555-25GEA
73
EUR
TCIAB1555-25G
TCIAB1555-5G
2-Bromo-4-fluorofenol ≥98.0% (by GC, titration analysis)
2-Bromo-4-fluorofenol
Formula:
C₆H₄BrFO MW: 191 g/mol Pto. Ebullición: 89°C |
Número MDL:
MFCD00010614 Núm. CAS: 496-69-5 |
Resultados test especificaciones
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 43.0 to 46.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...