3-Cloro-p-anísaldehído ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
3-Chloro-4-methoxybenzaldehyde
C1797-1G
C1797-5G
TCIAC1797-1GEA
35
EUR
TCIAC1797-1G
TCIAC1797-5G
3-Cloro-p-anísaldehído ≥98.0% (por GC)
3-Cloro-p-anísaldehído
Formula:
C₈H₇ClO₂ MW: 170,6 g/mol |
Número MDL:
MFCD00016601 Núm. CAS: 4903-09-7 EINECS: 225-532-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 56.0 to 60.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...