2-Cloro-4-nitroanísol ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-Chloro-2-methoxy-5-nitrobenzene
C1352-25G
C1352-5G
TCIAC1352-25GEA
115
EUR
TCIAC1352-25G
TCIAC1352-5G
2-Cloro-4-nitroanísol ≥98.0% (por GC)
2-Cloro-4-nitroanísol
Formula:
C₇H₆ClNO₃ |
Número MDL:
MFCD00060681 Núm. CAS: 4920-79-0 EINECS: 225-547-4 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 95.0 to 98.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...