Ácido 2,3,5,6-tetrafluorobenzoico ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

T1881-5G T1881-1G
TCIAT1881-5GEA 81 EUR
TCIAT1881-5G TCIAT1881-1G
Ácido 2,3,5,6-tetrafluorobenzoico ≥98.0% (by GC, titration analysis)
Ácido 2,3,5,6-tetrafluorobenzoico
Formula: C₇H₂F₄O₂
MW: 194,09 g/mol
Número MDL: MFCD00002408
Núm. CAS: 652-18-6
EINECS: 435-890-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4cfc563837cab1cbf78515fe1c2d61c8d22accb&vid=a4cfc563837cab1cbf78515fe1c2d61c8d22accb&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR