Ácido 2,3-difluorobenzoico ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
D2058-25G
D2058-5G
TCIAD2058-25GEA
174
EUR
TCIAD2058-25G
TCIAD2058-5G
Ácido 2,3-difluorobenzoico ≥98.0% (by GC, titration analysis)
Ácido 2,3-difluorobenzoico
Formula:
C₇H₄F₂O₂ MW: 158.1 g/mol |
Número MDL:
MFCD00010267 Núm. CAS: 4519-39-5 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 161.0 to 164.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...