3,4,5-Trifluoroanilina ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
T2355-1G
T2355-5G
TCIAT2355-1GEA
27
EUR
TCIAT2355-1G
TCIAT2355-5G
3,4,5-Trifluoroanilina ≥98.0% (por GC)
3,4,5-Trifluoroanilina
Formula:
C₆H₄F₃N MW: 147,1 g/mol |
Número MDL:
MFCD00083522 Núm. CAS: 163733-96-8 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...