2-Amino-4-clorobenzotiazol ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Clorobenzotiazol-2-ilamina
A1087-25G
A1087-5G
TCIAA1087-25GEA
333
EUR
TCIAA1087-25G
TCIAA1087-5G
2-Amino-4-clorobenzotiazol ≥98.0% (by HPLC)
2-Amino-4-clorobenzotiazol
Formula:
C₇H₅ClN₂S MW: 184,65 g/mol |
Número MDL:
MFCD00005791 Núm. CAS: 19952-47-7 EINECS: 243-439-5 |
Resultados test especificaciones
Appearance | White to Amber powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 202.0 to 207.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...