3-(2-Aminoetil)-5-metoxiindol

Proveedor: TCI
Atención

M0131-1G M0131-100MG M0131-5G
TCIAM0131-1GEA 79 EUR
TCIAM0131-1G TCIAM0131-100MG TCIAM0131-5G
3-(2-Aminoetil)-5-metoxiindol
3-(2-Aminoetil)-5-metoxiindol
Formula: C₁₁H₁₄N₂O
MW: 190,24 g/mol
Número MDL: MFCD00005662
Núm. CAS: 608-07-1
EINECS: 210-153-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4b13f4e6e1475dc8e9b93f7a2585be22d27271b&vid=e4b13f4e6e1475dc8e9b93f7a2585be22d27271b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR