4-Benciloxiindol ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
B2811-1G
B2811-5G
TCIAB2811-1GEA
62
EUR
TCIAB2811-1G
TCIAB2811-5G
4-Benciloxiindol ≥98.0% (por GC)
4-Benciloxiindol
Formula:
C₁₅H₁₃NO |
Número MDL:
MFCD00047200 Núm. CAS: 20289-26-3 EINECS: 243-690-0 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...