Ácido 3-amino-4-bromobenzoico ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Amino-4-bromobenzoic Acid
A2441-25G
A2441-5G
TCIAA2441-25GEA
268
EUR
TCIAA2441-25G
TCIAA2441-5G
Ácido 3-amino-4-bromobenzoico ≥98.0% (by HPLC, titration analysis)
3-Amino-4-bromobenzoic acid
Formula:
C₇H₆BrNO₂ MW: 216,03 g/mol |
Número MDL:
MFCD00579106 Núm. CAS: 2840-29-1 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystalline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 226.0 to 230.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...