Ácido 4-cloro-3-sulfamoilbenzoico ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

A1189-25G A1189-500G
TCIAA1189-25GEA 21 EUR
TCIAA1189-25G TCIAA1189-500G
Ácido 4-cloro-3-sulfamoilbenzoico ≥98.0% (by HPLC, titration analysis)
Ácido 4-cloro-3-sulfamoilbenzoico
Formula: C₇H₆ClNO₄S
Número MDL: MFCD00012375
Núm. CAS: 1205-30-7
EINECS: 214-882-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f922364256e974358aa23dccd78d690bffd72ea&vid=7f922364256e974358aa23dccd78d690bffd72ea&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR