Cloruro de 4-cloroanilinio ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
C0115-25G
C0115-500G
TCIAC0115-25GEA
21
EUR
TCIAC0115-25G
TCIAC0115-500G
Cloruro de 4-cloroanilinio ≥98.0% (by titrimetric analysis)
Cloruro de 4-cloroanilinio
Formula:
C₆H₇Cl₂N Pto. de fusión: 240°C |
Número MDL:
MFCD00042057 Núm. CAS: 20265-96-7 EINECS: 243-656-5 UN: 2018 ADR: 6.1,II |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Water | within almost transparency |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...