o-Dianisidina ≥98.0% (by HPLC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
3,3'-Dimetoxibencidina
D1344-25G
D1344-100G
TCIAD1344-25GEA
33
EUR
TCIAD1344-25G
TCIAD1344-100G
o-Dianisidina ≥98.0% (by HPLC)
o-Dianisidina
Formula:
C₁₄H₁₆N₂O₂ MW: 244.29 g/mol |
Número MDL:
MFCD00008372 Núm. CAS: 119-90-4 EINECS: 204-355-4 UN: 2811 ADR: 6.1 III Índice Merck: 14,02991 |
|
Resultados test especificaciones
Appearance | Yellow to Amber to Dark purple powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 136.0 to 140.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...