1-Boc-4-piperidona ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone
B2365-25G
B2365-5G
TCIAB2365-25GEA
89
EUR
TCIAB2365-25G
TCIAB2365-5G
1-Boc-4-piperidona ≥98.0% (por GC)
1-Boc-4-piperidona
Formula:
C₁₀H₁₇NO₃ MW: 199.25 g/mol |
Número MDL:
MFCD00151800 Núm. CAS: 79099-07-3 |
|
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...