1-Feniloxindol ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
1-Phenyl-1,3-dihydro-2H-indol-2-one
P1769-25G
P1769-5G
TCIAP1769-25GEA
90
EUR
TCIAP1769-25G
TCIAP1769-5G
1-Feniloxindol ≥98.0% (por GC)
1-Feniloxindol
Formula:
C₁₄H₁₁NO MW: 209,25 g/mol |
Número MDL:
MFCD00234850 Núm. CAS: 3335-98-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 120.0 to 126.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...