Trifluorometanosulfonato de 1-butil-3-metilimidazolio ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: 1-Butyl-3-methylimidazolium Trifluoromethanesulphonate

B2337-25G B2337-5G
TCIAB2337-25GEA 221 EUR
TCIAB2337-25G TCIAB2337-5G
Trifluorometanosulfonato de 1-butil-3-metilimidazolio ≥98.0% (by total nitrogen basis)
Trifluorometanosulfonato de 1-butil-3-metilimidazolio
Formula: C₉H₁₅F₃N₂O₃S
MW: 288,29 g/mol
Número MDL: MFCD03427620
Núm. CAS: 174899-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d538885fe22f6291fa72b11cd961b328556ba4b3&vid=d538885fe22f6291fa72b11cd961b328556ba4b3&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Yellow to Green clear liquid
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR