2-(2-Hidroxifenil)benzotiazol ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Benzotiazol-2-ilfenol
H0973-25G
H0973-5G
TCIAH0973-25GEA
120
EUR
TCIAH0973-25G
TCIAH0973-5G
2-(2-Hidroxifenil)benzotiazol ≥98.0% (by HPLC, titration analysis)
2-(2-Hidroxifenil)benzotiazol
Formula:
C₁₃H₉NOS MW: 227,29 g/mol |
Número MDL:
MFCD00022869 Núm. CAS: 3411-95-8 EINECS: 222-299-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 129.0 to 133.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...