Ácido bifenilo-2,2'-dicarboxílico ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

D0864-25G D0864-100G
TCIAD0864-25GEA 173 EUR
TCIAD0864-25G TCIAD0864-100G
Ácido bifenilo-2,2'-dicarboxílico ≥98.0% (by GC, titration analysis)
Ácido bifenilo-2,2'-dicarboxílico
Formula: C₁₄H₁₀O₄
Número MDL: MFCD00002464
Núm. CAS: 482-05-3
EINECS: 207-576-4
Índice Merck: 14,03310
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99157a515a09b6bbc0b274f21bcc1e790f586cf1&vid=99157a515a09b6bbc0b274f21bcc1e790f586cf1&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 230.0 to 236.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR