2-Imino-1-metilimidazolidin-4-ona ≥99.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
C0398-25G
C0398-500G
C0398-100G
TCIAC0398-25GEA
30
EUR
TCIAC0398-25G
TCIAC0398-500G
TCIAC0398-100G
2-Imino-1-metilimidazolidin-4-ona ≥99.0% (by HPLC, titration analysis)
2-Imino-1-metilimidazolidin-4-ona
Formula:
C₄H₇N₃O |
Número MDL:
MFCD00059730 Núm. CAS: 60-27-5 EINECS: 200-466-7 Índice Merck: 14,02569 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...