Ácido 2-metil-2-pentenoico ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

M0784-25G M0784-5G
TCIAM0784-25GEA 51 EUR
TCIAM0784-25G TCIAM0784-5G
Ácido 2-metil-2-pentenoico ≥98.0% (by GC, titration analysis)
Ácido 2-metil-2-pentenoico
Formula: C₆H₁₀O₂
Número MDL: MFCD00002655
Núm. CAS: 3142-72-1
EINECS: 221-552-0
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a736fa32ca68cce5cb9cab809aca619d6d2f6f2&vid=5a736fa32ca68cce5cb9cab809aca619d6d2f6f2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Freezing point 20.0 to 25.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR