α-Cloro-3-nitrotolueno ≥99.0% (por GC)

Proveedor: TCI

Sinónimos: 3-Nitrobencilo cloruro, m-Nitrobencilo cloruro

C0709-25G C0709-5G
TCIAC0709-25GEA 140 EUR
TCIAC0709-25G TCIAC0709-5G
α-Cloro-3-nitrotolueno ≥99.0% (por GC)
α-Cloro-3-nitrotolueno
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36c67f84043e8e60b3462df397a1bec8c024c5ca&vid=b1db3a93587bb14ec1efde30579c21769515f973&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 45.0 to 49.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR