α-α-α',α'-Tetrabromo-p-xileno ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
alpha,alpha,alpha',alpha'-Tetrabromo-p-xylene, p-Xylylene Tetrabromide, 1,4-Bis(dibromomethyl)benzene
T0053-25G
T0053-5G
TCIAT0053-25GEA
447
EUR
TCIAT0053-25G
TCIAT0053-5G
α-α-α',α'-Tetrabromo-p-xileno ≥98.0% (por GC)
α-α-α',α'-Tetrabromo-p-xileno
Formula:
C₈H₆Br₄ MW: 421.75 g/mol |
Número MDL:
MFCD00017857 Núm. CAS: 1592-31-0 EINECS: 216-473-3 UN: 1759 ADR: 8 III |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 168.0 to 172.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...