4-Aminobencilamina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
4-(Aminomethyl)aniline, alpha-Amino-p-toluidine
A1436-25G
A1436-250G
TCIAA1436-25GEA
111
EUR
TCIAA1436-25G
TCIAA1436-250G
4-Aminobencilamina ≥98.0% (by GC, titration analysis)
4-Aminobenzylamine
Formula:
C₇H₁₀N₂ MW: 122,17 g/mol |
Número MDL:
MFCD00075513 Núm. CAS: 4403-71-8 UN: 3259 ADR: 8,II |
Resultados test especificaciones
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...