1,3-Dietilurea ≥98.0% (by GC, total nitrogen)

Proveedor: TCI

D0534-25G D0534-500G
TCIAD0534-25GEA 40 EUR
TCIAD0534-25G TCIAD0534-500G
1,3-Dietilurea ≥98.0% (by GC, total nitrogen)
1,3-Dietilurea
Formula: C₅H₁₂N₂O
Número MDL: MFCD00009028
Núm. CAS: 623-76-7
EINECS: 210-811-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2aadab0fc31dbfde503361501b35c237959e91&vid=ec2aadab0fc31dbfde503361501b35c237959e91&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 110.0 to 113.0 deg-C
Solubility in Water within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR