(S)-(-)-4-Metoxi-α-metilbencilamina ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: (S)-1-(4-Methoxyphenyl)ethylamine, (S)-(-)-1-(4-Methoxyphenyl)ethylamine

M1512-25G M1512-5G
TCIAM1512-25GEA 198 EUR
TCIAM1512-25G TCIAM1512-5G
(S)-(-)-4-Metoxi-α-metilbencilamina ≥98.0% (by GC, titration analysis)
(S)-(-)-4-Metoxi-α-metilbencilamina
Formula: C₉H₁₃NO
MW: 151,21 g/mol
Número MDL: MFCD00671660
Núm. CAS: 41851-59-6
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e672e29519de0a6468626d0161581195570f77ac&vid=e672e29519de0a6468626d0161581195570f77ac&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 90.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR