N-Fenil-2,4-dinitroanilina ≥98.0% (por GC)

Proveedor: TCI
Atención

D0278-25G D0278-100G
TCIAD0278-25GEA 41 EUR
TCIAD0278-25G TCIAD0278-100G
N-Fenil-2,4-dinitroanilina ≥98.0% (por GC)
N-Fenil-2,4-dinitroanilina
Formula: C₁₂H₉N₃O₄
Número MDL: MFCD00007223
Núm. CAS: 961-68-2
EINECS: 213-508-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9acb4424a616e8f76705e5f4d802340660ade683&vid=9acb4424a616e8f76705e5f4d802340660ade683&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 157.0 to 160.0 deg-C
Solubility in hot Toluene within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR