Ácido 4-ciclohexilbenzoico ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

C1523-25G C1523-5G
TCIAC1523-25GEA 203 EUR
TCIAC1523-25G TCIAC1523-5G
Ácido 4-ciclohexilbenzoico ≥98.0% (by GC, titration analysis)
Ácido 4-ciclohexilbenzoico
Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Número MDL: MFCD00039459
Núm. CAS: 20029-52-1
EINECS: 243-472-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6824b38af8e8dd1653a9daccc0ab6042e319f8f&vid=344d9931a92d8d4d7eb87d7c3714dec77ad01536&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 196.0 to 201.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR