4-Cloropiridina clorhidrato ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

Sinónimos: 4-Chloropyridinium chloride

C0281-25G C0281-250G
TCIAC0281-25GEA 46 EUR
TCIAC0281-25G TCIAC0281-250G
4-Cloropiridina clorhidrato ≥98.0% (by HPLC, titration analysis)
4-Cloropiridina clorhidrato
Formula: C₅H₅Cl₂N
MW: 150.01 g/mol
Número MDL: MFCD00012829
Núm. CAS: 7379-35-3
EINECS: 230-946-1
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c315c6ffcea118609f377b883e0039af118863b&vid=8c315c6ffcea118609f377b883e0039af118863b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR