trans-2,5-Dimetilpiperazina ≥99.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

D0763-25G D0763-5G
TCIAD0763-25GEA 66 EUR
TCIAD0763-25G TCIAD0763-5G
trans-2,5-Dimetilpiperazina ≥99.0% (by GC, titration analysis)
trans-2,5-Dimetilpiperazina
Formula: C₆H₁₄N₂
MW: 114,19 g/mol
Número MDL: MFCD00070516
Núm. CAS: 2815-34-1
UN: 2926
ADR: 4.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d5d08b6d21fe152ee73a95ba6e38b0f9aac947c1&vid=d5d08b6d21fe152ee73a95ba6e38b0f9aac947c1&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 116.0 to 120.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR