Piridinio tribromuro ≥85.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide

P0825-25G P0825-500G P0825-100G
TCIAP0825-25GEA 20 EUR
TCIAP0825-25G TCIAP0825-500G TCIAP0825-100G
Piridinio tribromuro ≥85.0% (by titrimetric analysis)
Piridinio tribromuro
Formula: C₅H₆Br₃N
MW: 319.82 g/mol
Número MDL: MFCD00013223
Núm. CAS: 39416-48-3
EINECS: 254-446-8
UN: 1759
ADR: 8 III
Índice Merck: 14,07973
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34599efa161a53786b27ab6e6d85ee6a381f857b&vid=34599efa161a53786b27ab6e6d85ee6a381f857b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Iodometric Titration) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR