4-(Boc-amino)piridina ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

B3331-25G B3331-5G
TCIAB3331-25GEA 271 EUR
TCIAB3331-25G TCIAB3331-5G
4-(Boc-amino)piridina ≥98.0% (by GC, titration analysis)
4-(Boc-amino)piridina
Formula: C₁₀H₁₄N₂O₂
Número MDL: MFCD02179232
Núm. CAS: 98400-69-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6ece3176304370ee3360e61e92f5865f2c7fa18&vid=b6ece3176304370ee3360e61e92f5865f2c7fa18&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 148.0 to 152.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR