Tropina ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
endo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol
T0534-25G
T0534-5G
TCIAT0534-25GEA
88
EUR
TCIAT0534-25G
TCIAT0534-5G
Tropina ≥98.0% (by GC, titration analysis)
Tropina
Formula:
C₈H₁₅NO MW: 141,21 g/mol |
Número MDL:
MFCD00180667 Núm. CAS: 120-29-6 EINECS: 204-384-2 Índice Merck: 14,09781 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...