Ácido 3-hidroxi-2-piridincarboxílico ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Hydroxypyridine-2-carboxylic acid
H0787-25G
H0787-5G
TCIAH0787-25GEA
288
EUR
TCIAH0787-25G
TCIAH0787-5G
Ácido 3-hidroxi-2-piridincarboxílico ≥98.0% (by titrimetric analysis)
Ácido 3-hidroxi-2-piridincarboxílico
Formula:
C₆H₅NO₃ MW: 139,11 g/mol |
Número MDL:
MFCD00006294 Núm. CAS: 874-24-8 EINECS: 212-859-0 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in 1mol/L NaOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...