Ácido 5-fluoro-2-piridincarboxílico ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Fluoro-2-pyridinecarboxylic acid, 5-Fluoro-2-picolinic acid, 5-Fluoropyridine-2-carboxylic acid
F0838-1G
F0838-5G
TCIAF0838-1GEA
41
EUR
TCIAF0838-1G
TCIAF0838-5G
Ácido 5-fluoro-2-piridincarboxílico ≥98.0% (by GC, titration analysis)
Ácido 5-fluoro-2-piridincarboxílico
Formula:
C₆H₄FNO₂ MW: 141,1 g/mol |
Número MDL:
MFCD04114196 Núm. CAS: 107504-08-5 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 163.0 to 167.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...