DL-α-Fenilglicina ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Dl-A-Aminophenylacetic Acid, H-DL-Phg-OH, DL-2-Phenylglycine
P0326-25G
P0326-500G
TCIAP0326-25GEA
18
EUR
TCIAP0326-25G
TCIAP0326-500G
DL-α-Fenilglicina ≥98.0% (by HPLC, titration analysis)
DL-α-Fenilglicina
Formula:
C₈H₉NO₂ MW: 151,16 g/mol |
Número MDL:
MFCD00064402 Núm. CAS: 2835-06-5 EINECS: 220-608-1 Índice Merck: 14,07291 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Sodium hydroxide solution | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...