Bencenosulfinato de sodio dihidrato ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Benzenesulphinic acid sodium salt dihydrate
B0027-25G
B0027-500G
B0027-100G
TCIAB0027-25GEA
16
EUR
TCIAB0027-25G
TCIAB0027-500G
TCIAB0027-100G
Bencenosulfinato de sodio dihidrato ≥98.0% (by titrimetric analysis)
Bencenosulfinato de sodio dihidrato
Formula:
C₆H₅NaO₂S·2H₂O MW: 200.19 g/mol |
Número MDL:
MFCD00149638 Núm. CAS: 25932-11-0 EINECS: 212-842-8 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Sodium hypochlorite Method) | min. 98.0 % (after drying) |
Solubility in Water | almost transparency |
Drying loss | 16.0 to 20.0 % |
Benzene sulfonic acid | max. 0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...