(4-Hidroxifenil)acetamida ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
H0853-25G
H0853-500G
TCIAH0853-25GEA
25
EUR
TCIAH0853-25G
TCIAH0853-500G
(4-Hidroxifenil)acetamida ≥98.0% (by HPLC, total nitrogen)
(4-Hidroxifenil)acetamida
Formula:
C₈H₉NO₂ |
Número MDL:
MFCD00017145 Núm. CAS: 17194-82-0 EINECS: 241-235-0 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 175.0 to 179.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...