Ácido (R)-(+)-2-[[(1-feniletil)amino]carbonil]benzoico ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
M1622-1G
M1622-5G
TCIAM1622-1GEA
41
EUR
TCIAM1622-1G
TCIAM1622-5G
Ácido (R)-(+)-2-[[(1-feniletil)amino]carbonil]benzoico ≥98.0% (by HPLC, titration analysis)
Ácido (R)-(+)-2-[[(1-feniletil)amino]carbonil]benzoico
Formula:
C₁₆H₁₅NO₃ |
Número MDL:
MFCD00063050 Núm. CAS: 21752-35-2 EINECS: 244-570-0 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 126.0 to 130.0 deg-C |
Specific rotation [a]20/D | +45.0 to +48.0 deg (C=2,EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...