(±)-Ácido 2-phenoxypropionico ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
2-Phenoxypropanoic acid, 2-Phenoxypropionic acid, alpha-Methylphenoxyacetic acid
P0318-25G
P0318-500G
TCIAP0318-25GEA
16
EUR
TCIAP0318-25G
TCIAP0318-500G
(±)-Ácido 2-phenoxypropionico ≥98.0% (by GC, titration analysis)
(±)-Ácido 2-phenoxypropionico
Formula:
C₉H₁₀O₃ MW: 166,18 g/mol |
Número MDL:
MFCD00002643 Núm. CAS: 940-31-8 EINECS: 213-370-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...