Acetato de formamidina ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

F0154-25G F0154-250G
TCIAF0154-25GEA 19 EUR
TCIAF0154-25G TCIAF0154-250G
Acetato de formamidina ≥98.0% (by titrimetric analysis)
Acetato de formamidina
Formula: C₃H₈N₂O₂
MW: 104,11 g/mol
Número MDL: MFCD00012866
Núm. CAS: 3473-63-0
EINECS: 222-442-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=464974b91ea758db28e562a2ab49f360a6953160&vid=464974b91ea758db28e562a2ab49f360a6953160&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR