Ácido 3-(4-fluorobenzoil)propionico ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

F0379-25G F0379-5G
TCIAF0379-25GEA 250 EUR
TCIAF0379-25G TCIAF0379-5G
Ácido 3-(4-fluorobenzoil)propionico ≥97.0% (by GC, titration analysis)
3-(4-Fluorobenzoyl)propionic acid
Formula: C₁₀H₉FO₃
MW: 196,18 g/mol
Número MDL: MFCD00002793
Núm. CAS: 366-77-8
EINECS: 206-679-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5867135c3ca306d73bcf0a0b375bebc72c3460d0&vid=5867135c3ca306d73bcf0a0b375bebc72c3460d0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 101.0 to 104.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR