4,6-Dicloro-5-metilpirimidina ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
D4091-25G
D4091-5G
TCIAD4091-25GEA
294
EUR
TCIAD4091-25G
TCIAD4091-5G
4,6-Dicloro-5-metilpirimidina ≥98.0% (por GC)
4,6-Dicloro-5-metilpirimidina
Formula:
C₅H₄Cl₂N₂ MW: 163,01 g/mol |
Número MDL:
MFCD01646112 Núm. CAS: 4316-97-6 ADR: 8,II |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...