3-Piperidinametanol ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
H0737-25G
H0737-5G
TCIAH0737-25GEA
78
EUR
TCIAH0737-25G
TCIAH0737-5G
3-Piperidinametanol ≥98.0% (by GC, titration analysis)
3-Piperidinametanol
Formula:
C₆H₁₃NO Pto. Ebullición: 107°C |
Número MDL:
MFCD00005997 Núm. CAS: 4606-65-9 EINECS: 225-013-0 UN: 3263 ADR: 8,III |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 59.0 to 64.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...