Fenilhidroquinona ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

D0584-25G D0584-5G
TCIAD0584-25GEA 889 EUR
TCIAD0584-25G TCIAD0584-5G
Fenilhidroquinona ≥98.0% (by GC, titration analysis)
Fenilhidroquinona
Formula: C₁₂H₁₀O₂
Número MDL: MFCD00002342
Núm. CAS: 1079-21-6
EINECS: 214-091-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=045e5395e4e0a6c499ccf2ce7db31534db18019c&vid=045e5395e4e0a6c499ccf2ce7db31534db18019c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 97.0 %
Purity (Cerium redox Method) min. 97.0 %
Melting point 99.0 to 103.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR