2,6-Dicloro-p-benzoquinona ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2,6-Dichloro-1,4-benzoquinone

D0344-1G D0344-5G
TCIAD0344-1GEA 29 EUR
TCIAD0344-1G TCIAD0344-5G
2,6-Dicloro-p-benzoquinona ≥98.0% (by GC, titration analysis)
2,6-Dicloro-p-benzoquinona
Formula: C₆H₂Cl₂O₂
MW: 176.99 g/mol
Número MDL: MFCD00037159
Núm. CAS: 697-91-6
EINECS: 211-810-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca7a7dd49b94e8dcb435456b5f0b5caeb700a99a&vid=ca7a7dd49b94e8dcb435456b5f0b5caeb700a99a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Amber powder to crystal
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %
Melting point 120.0 to 123.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR