Benzoato de denatonio ≥98.0% (by HPLC)

Proveedor: TCI
Atención

D2124-25G D2124-5G D2124-100G
TCIAD2124-25GEA 192 EUR
TCIAD2124-25G TCIAD2124-5G TCIAD2124-100G
Benzoato de denatonio ≥98.0% (by HPLC)
Benzoato de denatonio
Formula: C₂₈H₃₄N₂O₃
Número MDL: MFCD00031578
Núm. CAS: 3734-33-6
EINECS: 223-095-2
Índice Merck: 14,02891
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f878a7c74820484d115936c53149013f34bfaaf0&vid=f878a7c74820484d115936c53149013f34bfaaf0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 % (Denatonium + Benzoic acid)
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 160.0 to 164.0 deg-C
Solubility in hot Water within very faint turbidity

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR