2,2,6,6-Tetrametil-4-piperidona ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Triacetonamine, 2,2,6,6-Tetramethylpiperidin-4-one
T1424-25G
T1424-500G
T1424-100G
TCIAT1424-25GEA
18
EUR
TCIAT1424-25G
TCIAT1424-500G
TCIAT1424-100G
2,2,6,6-Tetrametil-4-piperidona ≥98.0% (by GC, titration analysis)
2,2,6,6-Tetrametil-4-piperidona
Formula:
C₉H₁₇NO MW: 155,24 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00149393 Núm. CAS: 826-36-8 EINECS: 212-554-2 |
Resultados test especificaciones
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...