N-(2-(5-Metoxiindol-3-il)etil)acetamida ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
M1105-1G
M1105-5G
TCIAM1105-1GEA
16
EUR
TCIAM1105-1G
TCIAM1105-5G
N-(2-(5-Metoxiindol-3-il)etil)acetamida ≥98.0% (by HPLC, total nitrogen)
N-(2-(5-Metoxiindol-3-il)etil)acetamida
Formula:
C₁₃H₁₆N₂O₂ MW: 232.28 g/mol |
Número MDL:
MFCD00005655 Núm. CAS: 73-31-4 EINECS: 200-797-7 Índice Merck: 14,05816 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 117.0 to 120.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...