2-Amino-4-cloro-6-metilpirimidina ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Chloro-6-methylpyrimidin-2-amine

A1088-25G A1088-5G
TCIAA1088-25GEA 65 EUR
TCIAA1088-25G TCIAA1088-5G
2-Amino-4-cloro-6-metilpirimidina ≥98.0% (by titrimetric analysis)
2-Amino-4-chloro-6-methylpyrimidine
Formula: C₅H₆ClN₃
MW: 143,58 g/mol
Número MDL: MFCD00006091
Núm. CAS: 5600-21-5
EINECS: 227-018-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfec93e9596ad6b99f2c56a4cf53766363b74123&vid=bfec93e9596ad6b99f2c56a4cf53766363b74123&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 184.0 to 187.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR