Violeta de catecol

Proveedor: TCI

Sinónimos: Pyrocatechol Violet

P0568-1G P0568-5G
TCIAP0568-1GEA 16 EUR
TCIAP0568-1G TCIAP0568-5G
Violeta de catecol
Violeta de catecol
Formula: C₁₉H₁₄O₇S
MW: 386.38 g/mol
Número MDL: MFCD00005868
Núm. CAS: 115-41-3
EINECS: 204-088-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ccecb00b27d614e8c973b429a60fc5a574a3d02&vid=0ccecb00b27d614e8c973b429a60fc5a574a3d02&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Dark green to Dark red to Black powder to crystal
Lambda max. 442.0 to 445.0 nm (MeOH)
Absorbance (E1%1cm) min. 400 (MeOH, 442.0 to 445.0 nm) (calcd.on dried substance)
Drying loss max. 10.0 %
Sensitiveness Colorimetric Test : Turn Blue green with Cobalt and turn Purple by adding EDTA 2Na


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR